5-Amino-4,6-dichloropyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21030G100 AS21030G25
ACTIAS21030G100EA 0 GBP
ACTIAS21030G100 ACTIAS21030G25
5-Amino-4,6-dichloropyrimidine
5-Amino-4,6-dichloropyrimidine
Formula: C₄H₃Cl₂N₃
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 147 °C
MDL Number: MFCD00006108
CAS Number: 5413-85-4
EINECS: 226-503-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR