5-(2,6-Dichlorophenyl)-1H-tetrazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23762G1 AS23762G5
ACTIAS23762G1EA 0 GBP
ACTIAS23762G1 ACTIAS23762G5
5-(2,6-Dichlorophenyl)-1H-tetrazole
5-(2,6-Dichlorophenyl)-1H-tetrazole
Formula: C₇H₄Cl₂N₄
MDL Number: MFCD00798373
CAS Number: 50907-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR