4-Bromo-3,5-dimethylisoxazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS25302G100
AS25302G25
AS25302G5
ACTIAS25302G100EA
109
GBP
ACTIAS25302G100
ACTIAS25302G25
ACTIAS25302G5
4-Bromo-3,5-dimethylisoxazole
4-Bromo-3,5-dimethylisoxazole
Formula:
C₅H₆BrNO MW: 176.01 g/mol |
MDL Number:
MFCD00068187 CAS Number: 10558-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...