4,6-Dihydroxy-5-nitropyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Hydroxy-5-nitro-1H-pyrimidin-4-one, 4,6-Dihydroxy-5-nitropyrimdine, 5-Nitropyrimidine-4,6-diol

AS20827G100 AS20827G25
ACTIAS20827G100EA 0 GBP
ACTIAS20827G100 ACTIAS20827G25
4,6-Dihydroxy-5-nitropyrimidine
4,6-Dihydroxy-5-nitropyrimidine
Formula: C₄H₃N₃O₄
MW: 157.09 g/mol
Melting Pt: 300
MDL Number: MFCD00006110
CAS Number: 2164-83-2
EINECS: 218-504-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR