4,6-Dibromopyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS30581G1 AS30581G5
ACTIAS30581G1EA 0 GBP
ACTIAS30581G1 ACTIAS30581G5
4,6-Dibromopyrimidine
4,6-Dibromopyrimidine
Formula: C₄H₂Br₂N₂
MW: 237.88 g/mol
MDL Number: MFCD00233946
CAS Number: 36847-10-6
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR