4,4-Dimethylcyclohexanone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4-Dimethylcyclohexan-1-one

AS21407G1 AS21407G25 AS21407G5
ACTIAS21407G1EA 0 GBP
ACTIAS21407G1 ACTIAS21407G25 ACTIAS21407G5
4,4-Dimethylcyclohexanone
4,4-Dimethylcyclohexanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
MDL Number: MFCD00234965
CAS Number: 4255-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR