1-(4-Pyridyl)piperazine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(1-Piperazinyl)pyridine, 1-(Pyridin-4-yl)piperazine
AS20285G25
AS20285G5
ACTIAS20285G25EA
0
GBP
ACTIAS20285G25
ACTIAS20285G5
1-(4-Pyridyl)piperazine
1-(4-Pyridyl)piperazine
Formula:
C₉H₁₃N₃ MW: 163.22 g/mol Boiling Pt: 195…200 °C (0,3 mmHg) Melting Pt: 140 °C |
MDL Number:
MFCD00040745 CAS Number: 1008-91-9 EINECS: 213-764-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...