Thiophene-3-carbaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde

AS20035G100 AS20035G25
ACTIAS20035G100EA 0 GBP
ACTIAS20035G100 ACTIAS20035G25
Thiophene-3-carbaldehyde
Thiophene-3-carbaldehyde
Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1,28 g/cm³ (24 °C)
MDL Number: MFCD00005466
CAS Number: 498-62-4
EINECS: 207-865-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR