5-Fluoro-2-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20160G100 AS20160G25
ACTIAS20160G100EA 0 GBP
ACTIAS20160G100 ACTIAS20160G25
5-Fluoro-2-nitrobenzoic acid
5-Fluoro-2-nitrobenzoic acid
Formula: C₇H₄FNO₄
MDL Number: MFCD00055635
CAS Number: 320-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR