3-Chloropyridine-2-carbonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2080G25
AS2080G5
ACTIAS2080G25EA
0
GBP
ACTIAS2080G25
ACTIAS2080G5
3-Chloropyridine-2-carbonitrile
3-Chloropyridine-2-carbonitrile
Formula:
C₆H₃ClN₂ MW: 138.56 g/mol |
MDL Number:
MFCD03788834 CAS Number: 38180-46-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...