5-Bromonicotinonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromopyridine-3-carbonitrile, 3-Bromopyridine-5-carbonitrile, 5-Bromo-3-cyanopyridine, 3-Bromo-5-cyanopyridine

AS22754G1 AS22754G25 AS22754G5
ACTIAS22754G1EA 0 GBP
ACTIAS22754G1 ACTIAS22754G25 ACTIAS22754G5
5-Bromonicotinonitrile
5-Bromonicotinonitrile
Formula: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD00174363
CAS Number: 35590-37-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR