3-Bromopyridine-2-carbonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Bromopicolinonitrile, 3-Bromo-2-cyanopyridine
AS21017G1
AS21017G25
AS21017G5
ACTIAS21017G1EA
0
GBP
ACTIAS21017G1
ACTIAS21017G25
ACTIAS21017G5
3-Bromopyridine-2-carbonitrile
3-Bromopyridine-2-carbonitrile
Formula:
C₆H₃BrN₂ MW: 183.01 g/mol |
MDL Number:
MFCD02683288 CAS Number: 55758-02-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...