3-Amino-5-tert-butylisoxazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21767G25
AS21767G5
ACTIAS21767G25EA
0
GBP
ACTIAS21767G25
ACTIAS21767G5
3-Amino-5-tert-butylisoxazole
3-Amino-5-tert-butylisoxazole
Formula:
C₇H₁₂N₂O |
MDL Number:
MFCD00055620 CAS Number: 55809-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...