3-Amino-2-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methoxypyridin-3-amine, 2-Methoxy-3-pyridylamine

AS23108G25 AS23108G5
ACTIAS23108G25EA 0 GBP
ACTIAS23108G25 ACTIAS23108G5
3-Amino-2-methoxypyridine
3-Amino-2-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD00833386
CAS Number: 20265-38-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR