3,5-Dimethyl-4-iodoisoxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21267G100 AS21267G25
ACTIAS21267G100EA 0 GBP
ACTIAS21267G100 ACTIAS21267G25
3,5-Dimethyl-4-iodoisoxazole
3,5-Dimethyl-4-iodoisoxazole
Formula: C₅H₆INO
MDL Number: MFCD00173743
CAS Number: 10557-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR