Methyl 3,4-dihydroxybenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dihydroxybenzoic acid methyl ester, Methyl 3,4-dihydroxybenzoate

AS24694G100 AS24694G25
ACTIAS24694G100EA 81 GBP
ACTIAS24694G100 ACTIAS24694G25
Methyl 3,4-dihydroxybenzoate
Methyl 3,4-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00016437
CAS Number: 2150-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR