Ethyl-3,4-dihydroxybenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dihydroxybenzoic acid ethyl ester, Protocatechuic acid ethyl ester, Ethyl 3,4-dihydroxybenzoate

AS25239G100 AS25239G25
ACTIAS25239G100EA 0 GBP
ACTIAS25239G100 ACTIAS25239G25
Ethyl-3,4-dihydroxybenzoate
Ethyl 3,4-dihydroxybenzoate
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 134 °C
MDL Number: MFCD00002199
CAS Number: 3943-89-3
EINECS: 223-529-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR