3-(4-Fluorophenyl)propionic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22251G100
AS22251G25
AS22251G5
ACTIAS22251G100EA
303
GBP
ACTIAS22251G100
ACTIAS22251G25
ACTIAS22251G5
3-(4-Fluorophenyl)propionic acid
3-(4-Fluorophenyl)propionic acid
Formula:
C₉H₉FO₂ MW: 168.17 g/mol |
MDL Number:
MFCD00060327 CAS Number: 459-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...