3-(2-Fluorophenyl)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24705G25 AS24705G5
ACTIAS24705G25EA 0 GBP
ACTIAS24705G25 ACTIAS24705G5
3-(2-Fluorophenyl)propionic acid
3-(2-Fluorophenyl)propionic acid
Formula: C₉H₉FO₂
MDL Number: MFCD01310820
CAS Number: 1643-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR