2-Fluoro-5-(trifluoromethyl)benzaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20617G100
AS20617G25
ACTIAS20617G100EA
218
GBP
ACTIAS20617G100
ACTIAS20617G25
2-Fluoro-5-(trifluoromethyl)benzaldehyde
2-Fluoro-5-(trifluoromethyl)benzaldehyde
Formula:
C₈H₄F₄O MW: 192.11 g/mol |
MDL Number:
MFCD00061311 CAS Number: 146137-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...