2-Chloro-5-(trifluoromethyl)benzaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20544G100
AS20544G25
AS20544G5
ACTIAS20544G100EA
0
GBP
ACTIAS20544G100
ACTIAS20544G25
ACTIAS20544G5
2-Chloro-5-(trifluoromethyl)benzaldehyde
2-Chloro-5-(trifluoromethyl)benzaldehyde
Formula:
C₈H₄ClF₃O MW: 208.57 g/mol |
MDL Number:
MFCD00134430 CAS Number: 82386-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...