2-Amino-6-(trifluoromethyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20320G100
AS20320G25
AS20320G5
ACTIAS20320G100EA
0
GBP
ACTIAS20320G100
ACTIAS20320G25
ACTIAS20320G5
2-Amino-6-(trifluoromethyl)pyridine
2-Amino-6-(trifluoromethyl)pyridine
Formula:
C₆H₅F₃N₂ Boiling Pt: 200 °C (1013 hPa) Melting Pt: 78…80 °C |
MDL Number:
MFCD03093868 CAS Number: 34486-24-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...