2-Amino-5-bromopyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20786G100 AS20786G25
ACTIAS20786G100EA 0 GBP
ACTIAS20786G100 ACTIAS20786G25
2-Amino-5-bromopyrimidine
2-Amino-5-bromopyrimidine
Formula: C₄H₄BrN₃
MDL Number: MFCD00012341
CAS Number: 7752-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR