2-Amino-3-(trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20447G25 AS20447G5
ACTIAS20447G25EA 0 GBP
ACTIAS20447G25 ACTIAS20447G5
2-Amino-3-(trifluoromethyl)pyridine
2-Amino-3-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MDL Number: MFCD04972538
CAS Number: 183610-70-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR