2-Amino-1,3-propanediol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol
AS20932G100
AS20932G25
ACTIAS20932G100EA
0
GBP
ACTIAS20932G100
ACTIAS20932G25
2-Amino-1,3-propanediol
2-Amino-1,3-propanediol
Formula:
C₃H₉NO₂ Boiling Pt: 267 °C (1013 hPa) Melting Pt: 91…95 °C |
MDL Number:
MFCD00051487 CAS Number: 534-03-2 EINECS: 208-584-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...