2,6-Dibromonaphthalene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24285G25 AS24285G5
ACTIAS24285G25EA 0 GBP
ACTIAS24285G25 ACTIAS24285G5
2,6-Dibromonaphthalene
2,6-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MDL Number: MFCD01026462
CAS Number: 13720-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR