2,5-Difluoropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21781G25 AS21781G5
ACTIAS21781G25EA 0 GBP
ACTIAS21781G25 ACTIAS21781G5
2,5-Difluoropyridine
2,5-Difluoropyridine
Formula: C₅H₃F₂N
MDL Number: MFCD04114191
CAS Number: 84476-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR