Methyl-2,4-dihydroxybenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Dihydroxybenzoic acid methyl ester, Methyl 2,4-dihydroxybenzoate

AS24624G100 AS24624G25
ACTIAS24624G100EA 0 GBP
ACTIAS24624G100 ACTIAS24624G25
Methyl-2,4-dihydroxybenzoate
Methyl 2,4-dihydroxybenzoate
Formula: C₈H₈O₄
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 214…215 °C
MDL Number: MFCD00002276
CAS Number: 2150-47-2
EINECS: 218-428-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR