6-(Trifluoromethyl)uracil

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Trifluoromethyl)uracil, 2,4-Dihydroxy-6-trifluoromethylpyrimidine

AS21443G25 AS21443G5
ACTIAS21443G25EA 241 GBP
ACTIAS21443G25 ACTIAS21443G5
6-(Trifluoromethyl)uracil
6-(Trifluoromethyl)uracil
Formula: C₅H₃F₃N₂O₂
MW: 180.09 g/mol
MDL Number: MFCD01011762
CAS Number: 672-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR