1,3-Dimethyluracil

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dimethylpyrimidine-2,4(1H,3H)-dione

AS21015G100 AS21015G25
ACTIAS21015G100EA 77 GBP
ACTIAS21015G100 ACTIAS21015G25
1,3-Dimethyluracil
1,3-Dimethyluracil
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD00038065
CAS Number: 874-14-6
EINECS: 212-856-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR