1-Methyl-3-phenylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: MEFPIP

AS25356G100 AS25356G25 AS25356G5
ACTIAS25356G100EA 0 GBP
ACTIAS25356G100 ACTIAS25356G25 ACTIAS25356G5
1-Methyl-3-phenylpiperazine
1-Methyl-3-phenylpiperazine
Formula: C₁₁H₁₆N₂
MDL Number: MFCD03411603
CAS Number: 5271-27-2
EINECS: 431-180-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR