4-Fluoro-3-(trifluoromethyl)phenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-5-hydroxybenzotrifluoride, 4-Fluoro-3-trifluoromethylphenol

AS23325G100 AS23325G25
ACTIAS23325G100EA 0 GBP
ACTIAS23325G100 ACTIAS23325G25
4-Fluoro-3-(trifluoromethyl)phenol
4-Fluoro-3-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 84 °C (1013 hPa)
MDL Number: MFCD00043875
CAS Number: 61721-07-1
EINECS: 432-560-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR