3,5-Bis(trifluoromethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24912G100 AS24912G25
ACTIAS24912G100EA 0 GBP
ACTIAS24912G100 ACTIAS24912G25
3,5-Bis(trifluoromethyl)benzonitrile
3,5-Bis(trifluoromethyl)benzonitrile
Formula: C₉H₃F₆N
MW: 239.12 g/mol
Boiling Pt: 155
Melting Pt: 16
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00000379
CAS Number: 27126-93-8
EINECS: 248-240-7
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR