1-Aminopyridinium iodide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Aminopyridinium Iodide, 1-Aminopyridine hydroiodide, 1-Aminopyridine HI

AS23072G100 AS23072G25
ACTIAS23072G100EA 0 GBP
ACTIAS23072G100 ACTIAS23072G25
1-Aminopyridinium iodide
1-Aminopyridinium iodide
Formula: C₅H₇IN₂
MW: 222.03 g/mol
Melting Pt: 161…162 °C
MDL Number: MFCD00051893
CAS Number: 6295-87-0
EINECS: 228-566-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR