2-Nitro-4-(trifluoromethoxy)aniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24810G100 AS24810G25
ACTIAS24810G100EA 176 GBP
ACTIAS24810G100 ACTIAS24810G25
2-Nitro-4-(trifluoromethoxy)aniline
2-Nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₅F₃N₂O₃
MW: 222.12 g/mol
MDL Number: MFCD00042326
CAS Number: 2267-23-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR