2,5-Dichloro-α-methylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21674G100 AS21674G25 AS21674G5
ACTIAS21674G100EA 0 GBP
ACTIAS21674G100 ACTIAS21674G25 ACTIAS21674G5
2,5-Dichloro-α-methylbenzyl alcohol
2,5-Dichloro-α-methylbenzyl alcohol
Formula: C₈H₈Cl₂O
MW: 191.06 g/mol
MDL Number: MFCD00021861
CAS Number: 1475-12-3
EINECS: 216-018-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR