3,5-Dibromoanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dibromo-5-methoxybenzene

AS25464G100 AS25464G25 AS25464G5
ACTIAS25464G100EA 177 GBP
ACTIAS25464G100 ACTIAS25464G25 ACTIAS25464G5
3,5-Dibromoanisole
3,5-Dibromoanisole
Formula: C₇H₆Br₂O
MW: 265.93 g/mol
MDL Number: MFCD02258848
CAS Number: 74137-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR