(R)-(-)-Ethyl mandelate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25081G100 AS25081G25
ACTIAS25081G100EA 0 GBP
ACTIAS25081G100 ACTIAS25081G25
(R)-(-)-Ethyl mandelate
(R)-(-)-Ethyl mandelate
Formula: C₁₀H₁₂O₃
MDL Number: MFCD00064248
CAS Number: 10606-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR