DL-Homoserine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
H-DL-HoSer-OH, H-DL-Hser-OH, H-DL-HomoSer-OH, (±)-Homoserine
AS22075G25
AS22075G5
ACTIAS22075G25EA
0
GBP
ACTIAS22075G25
ACTIAS22075G5
DL-Homoserine
DL-Homoserine
Formula:
C₄H₉NO₃ Boiling Pt: 276 °C (1013 hPa) Melting Pt: 151 °C |
MDL Number:
MFCD00002618 CAS Number: 1927-25-9 EINECS: 217-661-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...