o,α-Dimethylbenzyl alcohol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(1-Hydroxyethyl)toluene, 2-Methylphenyl methyl carbinol, α,2-Dimethylbenzyl alcohol, 1-(2-Methylphenyl)ethanol, 1-(o-Tolyl)ethanol
AS21669G100
AS21669G25
ACTIAS21669G100EA
428
GBP
ACTIAS21669G100
ACTIAS21669G25
o,α-Dimethylbenzyl alcohol
o,α-Dimethylbenzyl alcohol
Formula:
C₉H₁₂O MW: 136.19 g/mol |
MDL Number:
MFCD00046632 CAS Number: 7287-82-3 EINECS: 230-716-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...