2-Fluoro-α-methylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21668G100 AS21668G25 AS21668G5
ACTIAS21668G100EA 464 GBP
ACTIAS21668G100 ACTIAS21668G25 ACTIAS21668G5
2-Fluoro-α-methylbenzyl alcohol
2-Fluoro-α-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD00014401
CAS Number: 445-26-1
EINECS: 207-155-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR