4-Chloro-α-methylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21672G100 AS21672G25
ACTIAS21672G100EA 0 GBP
ACTIAS21672G100 ACTIAS21672G25
4-Chloro-α-methylbenzyl alcohol
4-Chloro-α-methylbenzyl alcohol
Formula: C₈H₉ClO
Density: 1.17 g/cm³ (20 °C)
MDL Number: MFCD00041038
CAS Number: 3391-10-4
EINECS: 222-223-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR