2-Chloro-α-methylbenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21670G100 AS21670G25
ACTIAS21670G100EA 0 GBP
ACTIAS21670G100 ACTIAS21670G25
2-Chloro-α-methylbenzyl alcohol
2-Chloro-α-methylbenzyl alcohol
Formula: C₈H₉ClO
MDL Number: MFCD00041037
CAS Number: 13524-04-4
EINECS: 236-868-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR