3-Bromo-α-methylbenzyl alcohol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Bromophenyl methyl carbinol, m-Bromo-α-methylbenzyl alcohol, 1-(3-Bromophenyl)ethanol
AS21662G100
AS21662G25
ACTIAS21662G100EA
0
GBP
ACTIAS21662G100
ACTIAS21662G25
3-Bromo-α-methylbenzyl alcohol
3-Bromo-α-methylbenzyl alcohol
Formula:
C₈H₉BrO MW: 201.06 g/mol |
MDL Number:
MFCD00021863 CAS Number: 52780-14-0 EINECS: 258-177-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...