2-Bromo-4-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21316G1 AS21316G25 AS21316G5
ACTIAS21316G1EA 0 GBP
ACTIAS21316G1 ACTIAS21316G25 ACTIAS21316G5
2-Bromo-4-nitropyridine
2-Bromo-4-nitropyridine
Formula: C₅H₃BrN₂O₂
MDL Number: MFCD03701383
CAS Number: 6945-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR