2-Isobutyl-3-methoxypyrazine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24537G100
AS24537G25
AS24537G5
ACTIAS24537G100EA
452
GBP
ACTIAS24537G100
ACTIAS24537G25
ACTIAS24537G5
2-Isobutyl-3-methoxypyrazine
2-Isobutyl-3-methoxypyrazine
Formula:
C₉H₁₄N₂O MW: 166.22 g/mol Boiling Pt: 237 °C (1013 hPa) Melting Pt: 107 °C Density: 0.99 g/cm³ (20 °C) |
MDL Number:
MFCD00006128 CAS Number: 24683-00-9 EINECS: 246-402-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...