2-Aminopyrazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Aminopyrazine, Pyrazin-2-amine

AS20099G100 AS20099G25
ACTIAS20099G100EA 0 GBP
ACTIAS20099G100 ACTIAS20099G25
2-Aminopyrazine
2-Aminopyrazine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 203
Melting Pt: 117…120 °C
MDL Number: MFCD00006137
CAS Number: 5049-61-6
EINECS: 225-748-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR