Tetrapropylammonium perruthenate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: TPAP, Tetra-n-propylammonium perruthenate(VII)

AS20276G1 AS20276G5
ACTIAS20276G1EA 0 GBP
ACTIAS20276G1 ACTIAS20276G5
Tetrapropylammonium perruthenate
Tetrapropylammonium perruthenate
Formula: C₁₂H₂₈NO₄Ru
Melting Pt: 165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074914
CAS Number: 114615-82-6
UN: 1479
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR