1-(2-Hydroxyphenyl)piperazine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
o-(1-Piperazinyl)phenol, 2-(1-Piperazinyl)phenol
AS24297G100
AS24297G25
ACTIAS24297G100EA
0
GBP
ACTIAS24297G100
ACTIAS24297G25
1-(2-Hydroxyphenyl)piperazine
1-(2-Hydroxyphenyl)piperazine
Formula:
C₁₀H₁₄N₂O Boiling Pt: 318 °C (1013 hPa) Melting Pt: 127 °C |
MDL Number:
MFCD00190246 CAS Number: 1011-17-2 EINECS: 213-782-5 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...