1-(4-Nitrophenyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22132G1 AS22132G5
ACTIAS22132G1EA 0 GBP
ACTIAS22132G1 ACTIAS22132G5
1-(4-Nitrophenyl)piperazine
1-(4-Nitrophenyl)piperazine
Formula: C₁₀H₁₃N₃O₂
Melting Pt: 136.85 °C
MDL Number: MFCD00005961
CAS Number: 6269-89-2
EINECS: 228-443-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR